missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

£38.30 - £123.00

Chemical Identifiers

CAS 182163-96-8
Molecular Formula C9H13BO5
Molecular Weight (g/mol) 212.008
MDL Number MFCD01075676
InChI Key RULQUTYJXDLRFL-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
PubChem CID 2734390
IUPAC Name (3,4,5-trimethoxyphenyl)boronic acid
SMILES B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11398186
View Documents
Thermo Scientific Alfa Aesar
L15191.03
1 g
£38.30
1g
Please to purchase this item. Need a web account? Register with us today!
11308196
View Documents
Thermo Scientific Alfa Aesar
L15191.06
5 g
£123.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

182163-96-8
212.008
RULQUTYJXDLRFL-UHFFFAOYSA-N
2734390
B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O
C9H13BO5
MFCD01075676
3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
(3,4,5-trimethoxyphenyl)boronic acid

Specifications

182163-96-8
C9H13BO5
8205051
RULQUTYJXDLRFL-UHFFFAOYSA-N
(3,4,5-trimethoxyphenyl)boronic acid
2734390
98%
∼245°C
MFCD01075676
3,4,5-trimethoxybenzeneboronic acid, 3,4,5-trimethoxyphenyl boronic acid, 3,4,5-trimethoxyphenyl boranediol, 3,4,5-trimethoxylphenylboronic acid, boronic acid, 3,4,5-trimethoxyphenyl, pubchem7894, acmc-1c5iu, ksc174i9j, 3,4,5-trimethoxy phenylboronic acid
B(C1=CC(=C(C(=C1)OC)OC)OC)(O)O
212.008
212.01
3,4,5-Trimethoxybenzeneboronic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only