Learn More
3,4,5-Trimethoxybenzylamine, 96%, Thermo Scientific Chemicals
£51.80 - £173.00
Chemical Identifiers
CAS | 18638-99-8 |
---|---|
Molecular Formula | C10H15NO3 |
Molecular Weight (g/mol) | 197.234 |
MDL Number | MFCD00017150 |
InChI Key | YUPUSBMJCFBHAP-UHFFFAOYSA-N |
Synonym | 3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine |
PubChem CID | 87736 |
IUPAC Name | (3,4,5-trimethoxyphenyl)methanamine |
SMILES | COC1=CC(=CC(=C1OC)OC)CN |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11393077
|
Thermo Scientific Alfa Aesar
L13461.06 |
5 g |
£51.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11303087
|
Thermo Scientific Alfa Aesar
L13461.14 |
25 g |
£173.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4,5-Trimethoxybenzylamine is used in the synthesis of benzyl 3-deoxy-3-(3,4,5-trimethoxybenzylamino)-β-L-xylopyranoside. It is a important organic intermediate. It is used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Not miscible or difficult to mix in water.
Notes
It is sensitive to air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
Chemical Identifiers
18638-99-8 | |
197.234 | |
YUPUSBMJCFBHAP-UHFFFAOYSA-N | |
87736 | |
COC1=CC(=CC(=C1OC)OC)CN |
C10H15NO3 | |
MFCD00017150 | |
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine | |
(3,4,5-trimethoxyphenyl)methanamine |
Specifications
18638-99-8 | |
120°C to 122°C (0.05 mmHg) | |
C10H15NO3 | |
MFCD00017150 | |
1107570 | |
3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine | |
YUPUSBMJCFBHAP-UHFFFAOYSA-N | |
(3,4,5-trimethoxyphenyl)methanamine | |
87736 | |
96% |
1.155 | |
>110°C (230°F) | |
1.548 | |
UN2735 | |
Air Sensitive | |
Not miscible or difficult to mix in water. | |
COC1=CC(=CC(=C1OC)OC)CN | |
197.234 | |
197.24 | |
3,4,5-Trimethoxybenzylamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
missing translation for 'einecsNumber' : 242-468-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only