missing translation for 'onlineSavingsMsg'
Learn More

3,4'-Bis(trifluoromethyl)benzophenone, 99%, Thermo Scientific™

Product Code. 15438909
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21084-22-0
Molecular Formula C15H8F6O
Molecular Weight (g/mol) 318.218
MDL Number MFCD00013561
InChI Key HWBXNAPXNCBCQO-UHFFFAOYSA-N
Synonym 3,4'-bis trifluoromethyl benzophenone, 3,4'-di trifluoromethyl benzophenone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl methanone, 3-trifluoromethyl phenyl-4-trifluoromethyl phenyl methanone, methanone, 3-trifluoromethyl phenyl 4-trifluoromethyl phenyl, 3,4-bis trifluoromethyl benzophenone, benzophenone, 3,4'-bis trifluoromethyl
PubChem CID 736046
IUPAC Name [3-(trifluoromethyl)phenyl]-[4-(trifluoromethyl)phenyl]methanone
SMILES C1=CC(=CC(=C1)C(F)(F)F)C(=O)C2=CC=C(C=C2)C(F)(F)F

Specifications

Melting Point 93°C to 95°C
Quantity 25 g
Beilstein 3064970
Formula Weight 318.22
Percent Purity 99%
Chemical Name or Material 3,4'-Bis(trifluoromethyl)benzophenone
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'