missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorophenoxy)-1,2-propanediol, 99%

Product Code. 11493627
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
missing translation for 'unitSize'
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11493627

missing translation for 'mfr': Thermo Scientific Alfa Aesar A17386.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-29-0 | C9H11ClO3 | 202.634 g/mol

It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It inhibits IgE-mediated histamine release. 3-(4-Chlorophenoxy)-1,2-propanediol is also used as an antimycotic agent. It is a centrally acting muscle relaxant.

Solubility
Slightly soluble in water.

Notes
Store at 78-82°C. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-29-0
Molecular Formula C9H11ClO3
Molecular Weight (g/mol) 202.634
MDL Number MFCD00021990
InChI Key MXOAEAUPQDYUQM-UHFFFAOYSA-N
Synonym chlorphenesin, 3-4-chlorophenoxy-1,2-propanediol, 3-4-chlorophenoxy propane-1,2-diol, chlorophenesin, chlorphenesinum, adermykon, gecophen, demykon, mycil, chlorphenesine
PubChem CID 7697
ChEBI CHEBI:3642
IUPAC Name 3-(4-chlorophenoxy)propane-1,2-diol
SMILES C1=CC(=CC=C1OCC(CO)O)Cl

Specifications

Melting Point 78°C to 82°C
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 202.64
Percent Purity 99%
Chemical Name or Material 3-(4-Chlorophenoxy)-1,2-propanediol
compliance-icons
Product Identifier
  • 3-(4-Chlorophenoxy)-1,2-propanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Chlorphenesin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.