missing translation for 'onlineSavingsMsg'
Learn More

3-(4-chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10793367
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 116525-66-7
Molecular Formula C13H8ClNO2S2
Molecular Weight (g/mol) 309.78
MDL Number MFCD00112478
InChI Key CCQQTQYIJRVXIU-UHFFFAOYSA-N
Synonym 3-4-chlorophenyl-4-cyano-5-methylthio thiophene-2-carboxylic acid, 2-thiophenecarboxylicacid, 3-4-chlorophenyl-4-cyano-5-methylthio, 3-4-chlorophenyl-4-cyano-5-methylsulfanyl thiophene-2-carboxylic acid, maybridge1_004446, acmc-1bss2, 3-4-chlorophenyl-4-cyano-5-methylthio-2-thiophenecarboxylic acid, 3-4-chlorophenyl-4-cyano-5-methylsulfanyl-thiophene-2-carboxylic acid, 3-4-chlorophenyl-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid
PubChem CID 2735791
SMILES CSC1=C(C#N)C(=C(S1)C(O)=O)C1=CC=C(Cl)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
IUPAC Name 3-(4-chlorophenyl)-4-cyano-5-methylsulfanylthiophene-2-carboxylic acid
Formula Weight 309.8
Percent Purity 97%
Chemical Name or Material 3-(4-Chlorophenyl)-4-cyano-5-(methylthio)thiophene-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.