missing translation for 'onlineSavingsMsg'
Learn More

3,4-Diaminobenzoic acid, 97%

Product Code. 10518290 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10518290

Brand: Thermo Scientific Acros 112101000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-05-6 | C7H8N2O2 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-05-6
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007726
InChI Key HEMGYNNCNNODNX-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-diamino, 4-carboxyphenyldiamine, 3,4-diamino benzoic acid, 4-carboxy-o-phenylenediamine, 3,4-diaminobenzoicacid, acmc-209mxf, 3,4-diaminobenzoesaeure, 3,4 diaminobenzoic acid, 3,4-diaminobenzenoic acid, 3,4-diamino-benzoic acid
PubChem CID 69263
IUPAC Name 3,4-diaminobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 215.0°C
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula (H2N)2C6H3CO2H
Quantity 100 g
Beilstein 14, 450
Solubility Information Solubility in water: 2.2g/L (20°C).
Formula Weight 152.15
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 3, 4-Diaminobenzoic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.