Learn More
3,4-Diaminotoluene, 97%, Thermo Scientific Chemicals
£26.90 - £79.20
Chemical Identifiers
CAS | 496-72-0 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00007728 |
InChI Key | DGRGLKZMKWPMOH-UHFFFAOYSA-N |
Synonym | 3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene |
PubChem CID | 10332 |
IUPAC Name | 4-methylbenzene-1,2-diamine |
SMILES | CC1=CC(=C(C=C1)N)N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10316370
|
Thermo Scientific
112431000 |
100 g | Glass bottle |
£26.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10696354
|
Thermo Scientific Acros
112435000 |
500 g | Glass bottle |
£79.20
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
496-72-0 | |
122.17 | |
DGRGLKZMKWPMOH-UHFFFAOYSA-N | |
10332 | |
CC1=CC(=C(C=C1)N)N |
C7H10N2 | |
MFCD00007728 | |
3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene | |
4-methylbenzene-1,2-diamine |
Specifications
496-72-0 | |
100.0 | |
Brown to Brown-Gray | |
Authentic | |
C7H10N2 | |
MFCD00007728 | |
3,4-diaminotoluene, 3,4-toluenediamine, 1,2-benzenediamine, 4-methyl, 4-methyl-o-phenylenediamine, toluene-3,4-diamine, 4-methyl-1,2-phenylenediamine, 4-methyl-1,2-benzenediamine, 3,4-tolylenediamine, 3,4-toluylenediamine, 1,2-diamino-4-methylbenzene | |
DGRGLKZMKWPMOH-UHFFFAOYSA-N | |
4-methylbenzene-1,2-diamine | |
10332 | |
97% | |
3, 4-Diaminotoluene |
96.0 | |
83.0°C to 90.0°C | |
265.0°C | |
96% min. (HPLC) | |
CH3C6H3(NH2)2 | |
13, 148 | |
Solubility in water: 16g/L (20°C). | |
CC1=CC(=C(C=C1)N)N | |
122.17 | |
122.17 | |
Crystalline Chunks or Flakes |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes serious eye irritation.
Suspected of causing genetic defects.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if y
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 207-826-2
- 3,4-Diaminotoluene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Germ cell mutagenicity Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H341-Suspected of causing genetic defects.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only