missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals Product Code.: 11339539

3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals

Product Code. 11339539
50 g
Click to view available options
Quantity:
50 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11339539

Brand: Thermo Scientific Alfa Aesar A15199.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-76-1
Molecular Formula C6H5Cl2N
Molecular Weight (g/mol) 162.013
MDL Number MFCD00007768
InChI Key SDYWXFYBZPNOFX-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroanilineShow MoreShow Less
PubChem CID 7257
ChEBI CHEBI:16767
IUPAC Name 3,4-dichloroaniline
SMILES C1=CC(=C(C=C1N)Cl)Cl

Specifications

Melting Point 70°C to 73°C
Boiling Point 272°C
Flash Point 135°C (275°F)
Quantity 50 g
UN Number UN3442
Beilstein 636837
Merck Index 14,3054
Formula Weight 162.02
Percent Purity 98%
Chemical Name or Material 3,4-Dichloroaniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2933077A Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberA15199.30,A15199.18,A15199.0E,A15199.36
Lot Number10222153 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA15199.30,A15199.18,A15199.0E,A15199.36
Lot NumberJ4863A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15199.30,A15199.18,A15199.0E,A15199.36
Lot NumberFA021097 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15199.30,A15199.18,A15199.0E,A15199.36
Lot Number10141038 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA15199.30,A15199.18,A15199.0E,A15199.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.