Learn More
3,4-Dichloroaniline, 98%, Thermo Scientific Chemicals
£20.30 - £36.38
Chemical Identifiers
CAS | 95-76-1 |
---|---|
Molecular Formula | C6H5Cl2N |
Molecular Weight (g/mol) | 162.013 |
MDL Number | MFCD00007768 |
InChI Key | SDYWXFYBZPNOFX-UHFFFAOYSA-N |
Synonym | 3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline |
PubChem CID | 7257 |
ChEBI | CHEBI:16767 |
IUPAC Name | 3,4-dichloroaniline |
SMILES | C1=CC(=C(C=C1N)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11339539
|
Thermo Scientific Alfa Aesar
A15199.18 |
50 g |
£20.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11349539
|
Thermo Scientific Alfa Aesar
A15199.30 |
250 g |
£23.01
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11432218
|
Thermo Scientific Alfa Aesar
A15199.36 |
500 g |
£36.38
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-76-1 | |
162.013 | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
7257 | |
3,4-dichloroaniline |
C6H5Cl2N | |
MFCD00007768 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
CHEBI:16767 | |
C1=CC(=C(C=C1N)Cl)Cl |
Specifications
95-76-1 | |
272°C | |
C6H5Cl2N | |
UN3442 | |
14,3054 | |
SDYWXFYBZPNOFX-UHFFFAOYSA-N | |
3,4-dichloroaniline | |
7257 | |
162.02 | |
3,4-Dichloroaniline |
70°C to 73°C | |
135°C (275°F) | |
MFCD00007768 | |
636837 | |
3,4-dichlorobenzenamine, 3,4-dichloraniline, 3,4-dichloranilin, 4,5-dichloroaniline, benzenamine, 3,4-dichloro, 1-amino-3,4-dichlorobenzene, 3,4-dca, 3,4-dichlorophenylamine, aniline, 3,4-dichloro, m,p-dichloroaniline | |
C1=CC(=C(C=C1N)Cl)Cl | |
162.013 | |
CHEBI:16767 | |
98% |
Safety and Handling
GHS H Statement
H310-H330-H318-H302-H317
Fatal in contact with skin.
Fatal if inhaled.
Causes serious eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.
P261-P264b-P270-P271-P272-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P311-P312-P330-P333+P313-P361-P363-P501c
H301+H311+H331-H317-H318-H500
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID
missing translation for 'einecsNumber' : 202-448-4
missing translation for 'rtecsNumber' : BX2625000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only