missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzaldoxime, 98%, Thermo Scientific™

Product Code. 11378295
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5331-92-0
Molecular Formula C7H5Cl2NO
Molecular Weight (g/mol) 190.02
MDL Number MFCD00017592
InChI Key ROBIUDOANJUDHD-ONNFQVAWSA-N
Synonym 3,4-dichlorobenzaldoxime, 3,4-dichlorobenzaldehyde oxime, 1e-3,4-dichlorobenzaldehyde oxime, e-n-3,4-dichlorophenyl methylidene hydroxylamine, ne-n-3,4-dichlorophenyl methylidene hydroxylamine, 3,4-dichlorophenyl hydroxyimino methane, 3,4-dichlorobenzaldehyde oxime #, z-n-3,4-dichlorophenyl methylidene hydroxylamine
PubChem CID 5704835
IUPAC Name (NE)-N-[(3,4-dichlorophenyl)methylidene]hydroxylamine
SMILES O\N=C\C1=CC=C(Cl)C(Cl)=C1

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 2206792
Formula Weight 190.03
Percent Purity 98%
Chemical Name or Material 3,4-Dichlorobenzaldoxime
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'