Learn More
3,4-Dichlorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
£20.40 - £186.00
Chemical Identifiers
CAS | 151169-75-4 |
---|---|
Molecular Formula | C6H5BCl2O2 |
Molecular Weight (g/mol) | 190.81 |
MDL Number | MFCD01074646 |
InChI Key | JKIGHOARKAIPJI-UHFFFAOYSA-N |
Synonym | 3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 |
PubChem CID | 2734330 |
IUPAC Name | (3,4-dichlorophenyl)boronic acid |
SMILES | OB(O)C1=CC=C(Cl)C(Cl)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11445707
|
Thermo Scientific Alfa Aesar
B24292.03 |
1 g |
£20.40
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11455707
|
Thermo Scientific Alfa Aesar
B24292.06 |
5 g |
£58.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465707
|
Thermo Scientific Alfa Aesar
B24292.14 |
25 g |
£186.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151169-75-4 | |
190.81 | |
JKIGHOARKAIPJI-UHFFFAOYSA-N | |
2734330 | |
OB(O)C1=CC=C(Cl)C(Cl)=C1 |
C6H5BCl2O2 | |
MFCD01074646 | |
3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 | |
(3,4-dichlorophenyl)boronic acid |
Specifications
151169-75-4 | |
C6H5BCl2O2 | |
7369790 | |
JKIGHOARKAIPJI-UHFFFAOYSA-N | |
(3,4-dichlorophenyl)boronic acid | |
2734330 | |
97% |
270°C to 274°C | |
MFCD01074646 | |
3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 | |
OB(O)C1=CC=C(Cl)C(Cl)=C1 | |
190.81 | |
190.82 | |
3,4-Dichlorobenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only