missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorobenzyl mercaptan, 97%, Thermo Scientific Chemicals

Product Code. 11362066
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of substituted pyridazines. 3,4-Dichlorobenzyl mercaptan helps to improve the semiconductor morphologies.

Solubility
Difficult to mix in water.

Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36480-40-7
Molecular Formula C7H6Cl2S
Molecular Weight (g/mol) 193.085
MDL Number MFCD00039651
InChI Key CEGBRSQPRQXALB-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzyl mercaptan, 3,4-dichlorobenzenemethanethiol, 3,4-dichlorophenyl methanethiol, 3,4-dichlorotoluene-alpha-thiol, benzenemethanethiol, 3,4-dichloro, acmc-1aex5, d0z0bb, 3,4-dichlorobenzylmercaptan, 3,4-dichloro-alpha-toluenethiol, 3,4-dichloro-phenyl-methanethiol
PubChem CID 118986
IUPAC Name (3,4-dichlorophenyl)methanethiol
SMILES C1=CC(=C(C=C1CS)Cl)Cl

Specifications

Density 1.362
Boiling Point 152°C to 153°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.607
Quantity 1 g
UN Number UN2810
Beilstein 3237473
Sensitivity Air Sensitive
Solubility Information Difficult to mix in water.
Formula Weight 193.1
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzyl mercaptan
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'