Learn More
3,4-Dichloroisocoumarin, 98%, Thermo Scientific Chemicals
Description
Useful in a rapid staining method for identification of macrophages; counts by this method were confirmed by the more complex morphological criteria, by phagocytosis, and by the presence of Fc receptors. It is an inhibitor of Chymotrypsin and Trypsin. ,4 Dichloroisocoumarin also inhibits granzymes A, B, and H, cathepsin G, neutrophil elastase, proteinase and blocks apoptotic internucleosomal DNA cleavage in thymocytes without the involvement of endonucleases.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Useful in a rapid staining method for identification of macrophages; counts by this method were confirmed by the more complex morphological criteria, by phagocytosis, and by the presence of Fc receptors. It is an inhibitor of Chymot
Specifications
Specifications
CAS | 51050-59-0 |
Color | White |
Molecular Formula | C9H4Cl2O2 |
MDL Number | MFCD00036960 |
Quantity | 10 mg |
UN Number | UN2811 |
Beilstein | 6802625 |
Synonym | 3,4-dichloroisocoumarin, 3,4-dcl, 1h-2-benzopyran-1-one,3,4-dichloro, 3,4-dci, 3,4 dichloroisocoumarin, 3,4-dichloro-1h-isochromen-1-one, 3,4-dichloro-1h-2-benzopyran-1-one, 1h-2-benzopyran-1-one, 3,4-dichloro, chembl24983, 3,4-dichloro-2-benzopyran-1-one |
Solubility Information | Soluble in DMF or DMSO at a concentration of 10mM |
InChI Key | SUGXUUGGLDCZKB-UHFFFAOYSA-N |
Show More |
RUO – Research Use Only