Learn More
3,4-Dichlorophenylboronic acid, 97%, Thermo Scientific Chemicals
£63.30 - £203.00
Chemical Identifiers
CAS | 151169-75-4 |
---|---|
Molecular Formula | C6H5BCl2O2 |
Molecular Weight (g/mol) | 190.81 |
MDL Number | MFCD01074646 |
InChI Key | JKIGHOARKAIPJI-UHFFFAOYSA-N |
Synonym | 3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 |
PubChem CID | 2734330 |
IUPAC Name | (3,4-dichlorophenyl)boronic acid |
SMILES | OB(O)C1=CC=C(Cl)C(Cl)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10744363
|
Thermo Scientific Acros
359090050 |
5 g | Glass bottle |
£63.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10073907
|
Thermo Scientific Acros
359090250 |
25 g | Glass bottle |
£203.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
151169-75-4 | |
190.81 | |
JKIGHOARKAIPJI-UHFFFAOYSA-N | |
2734330 | |
OB(O)C1=CC=C(Cl)C(Cl)=C1 |
C6H5BCl2O2 | |
MFCD01074646 | |
3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 | |
(3,4-dichlorophenyl)boronic acid |
Specifications
151169-75-4 | |
100.0 | |
White | |
96% min. (HPLC) | |
MFCD01074646 | |
JKIGHOARKAIPJI-UHFFFAOYSA-N | |
(3,4-dichlorophenyl)boronic acid | |
2734330 | |
97% | |
3, 4-Dichlorophenylboronic acid |
96.0 | |
284.0°C to 286.0°C | |
Authentic | |
C6H5BCl2O2 | |
3,4-dichlorophenyl boronic acid, 3,4-dichlorobenzeneboronic acid, 3,4-dichlorophenylboronicacid, 3,4-dichlorophenyl boranediol, 3,4-dichlorophenylbornic acid, chembl20776, 3,4-dichloro benzene boronic acid, boronic acid, 3,4-dichlorophenyl, pubchem1812 | |
OB(O)C1=CC=C(Cl)C(Cl)=C1 | |
190.81 | |
190.82 | |
Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre
GHS Signal Word: Warning
- 3,4-Dichlorophenylboronic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only