missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluoro-DL-phenylalanine, 97%, Thermo Scientific™

Product Code. 15414987
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32133-36-1
Molecular Formula C9H9F2NO2
Molecular Weight (g/mol) 201.17
MDL Number MFCD00061307
InChI Key PRAWYXDDKCVZTL-UHFFFAOYNA-N
Synonym 2-amino-3-3,4-difluorophenyl propanoic acid, 3,4-difluoro-dl-phenylalanine, dl-3,4-difluorophenylalanine, 3,4-difluorophenylalanine, 2-amino-3-3,4-difluoro-phenyl-propionic acid, 2-azanyl-3-3,4-bis fluoranyl phenyl propanoic acid, h-dl-phe 3,4-f2-oh, pubchem8478, acmc-209gg4, 3,4-difluoro-dlphenylalanine
PubChem CID 2737044
IUPAC Name 2-amino-3-(3,4-difluorophenyl)propanoic acid
SMILES NC(CC1=CC(F)=C(F)C=C1)C(O)=O

Specifications

Melting Point 232°C to 234°C
Formula Weight 201.18
Percent Purity 97%
Quantity 250 mg
Chemical Name or Material 3,4-Difluoro-DL-phenylalanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'