missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenol, 98%, Thermo Scientific Chemicals

Product Code. 11414324
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2713-33-9
Molecular Formula C6H4F2O
Molecular Weight (g/mol) 130.09
MDL Number MFCD00010315
InChI Key BNPWVUJOPCGHIK-UHFFFAOYSA-N
Synonym phenol, 3,4-difluoro, 3,4-difluoro phenol, 3 4-difluorophenol, 3,4-difluoro-phenol, pubchem2315, acmc-1cmaj, 3,4-difluorophenol, ksc202q5j, bnpwvujopcghik-uhfffaoysa
PubChem CID 75927
IUPAC Name 3,4-difluorophenol
SMILES OC1=CC=C(F)C(F)=C1

Specifications

Melting Point 34°C to 38°C
Boiling Point 84°C to 86°C (20 mmHg)
Flash Point 57°C (134°F)
Odor Phenol-like
Quantity 25 g
UN Number UN2921
Beilstein 2081085
Formula Weight 130.09
Percent Purity 98%
Chemical Name or Material 3,4-Difluorophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'