missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylmagnesium bromide, Thermo Scientific Chemicals

Product Code. 10604755
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 90897-92-0, 109-99-9
CAS Min % 9.01, 86.49
CAS Max % 13.51, 90.99
Color Brown to Yellow
Density 96%5g/mL
Packaging AcroSeal™ Glass Bottle
Molecular Formula C6H3BrF2Mg
Linear Formula F2C6H3MgBr
MDL Number MFCD00672004
Quantity 100 mL
Synonym 3,4-difluorophenylmagnesium bromide, bromo 3,4-difluorophenyl magnesium, 3,4-difluorophenyl magnesium bromide, c6h3brf2m, wzhavygmkgpnfn-uhfffaoysa-m, magnesium,bromo 3,4-difluorophenyl, 3,4-difluorophenylmagnesium bromide, 0.50 m in 2-methf, 3,4-difluorophenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,4-difluorophenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
InChI Key WZHAVYGMKGPNFN-UHFFFAOYSA-M
SMILES FC1=CC=C([Mg]Br)C=C1F
IUPAC Name magnesium;1,2-difluorobenzene-5-ide;bromide
Molecular Weight (g/mol) 217.30
PubChem CID 2778383
Formula Weight 217.29
Physical Form Solution
Chemical Name or Material 3, 4-Difluorophenylmagnesium bromide, 0.5M solution in THF
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3,4-Difluorophenylmagnesium bromide, 0.5M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'