missing translation for 'onlineSavingsMsg'
Learn More

3,4-Difluorophenylzinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific™

Product Code. 15465365
Click to view available options
Quantity:
50 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 737797-28-3
Molecular Formula C6H3BrF2Zn
MDL Number MFCD02260154
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive; May form a precipitate
Synonym bromo 3,4-difluorophenyl zinc, 3,4-difluorophenylzinc bromide, 0.5m solution in thf, 3,4-difluorophenylzinc bromide 0.5 m in tetrahydrofuran, 3,4-difluorophenylzinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key RPYBEKIISMVWQQ-UHFFFAOYSA-M
SMILES C1=[C-]C=C(C(=C1)F)F.[Zn+]Br
IUPAC Name bromozinc(1+);1,2-difluorobenzene-5-ide
Molecular Weight (g/mol) 258.371
PubChem CID 24722621
Formula Weight 258.37
Chemical Name or Material 3,4-Difluorophenylzinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'