missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,4-benzoxazine, 97%, Thermo Scientific Chemicals

Product Code. 15426487
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

Solubility
Sparingly soluble in water.(0.26 g/L) (25°C),

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5735-53-5
Molecular Formula C8H9NO
Molecular Weight (g/mol) 135.166
MDL Number MFCD02181098
InChI Key YRLORWPBJZEGBX-UHFFFAOYSA-N
Synonym benzomorpholine, 3,4-dihydro-2h-benzo b 1,4 oxazine, 3,4-dihydro-2h-benzo 1,4 oxazine, 2h-1,4-benzoxazine, 3,4-dihydro, 2,3-dihydro-1,4-benzoxazine, akos bc-1149, 3,4-dihydro-2h-1,4-benzoxazin, pubchem22677, acmc-1amxt, 3,4-dihydro-2h-1,4-benzooxazine
PubChem CID 585096
IUPAC Name 3,4-dihydro-2H-1,4-benzoxazine
SMILES C1COC2=CC=CC=C2N1

Specifications

Boiling Point 85°C (1 mmHg)
Quantity 1 g
Solubility Information Sparingly soluble in water.(0.26g/L) (25°C),
Formula Weight 135.17
Percent Purity 97%
Chemical Name or Material 3,4-Dihydro-2H-1,4-benzoxazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'