missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dihydro-2H-pyran, 99%, Thermo Scientific Chemicals Product Code.: 11307815

3,4-Dihydro-2H-pyran, 99%, Thermo Scientific Chemicals

Product Code. 11307815
25 mL
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11307815

Brand: Thermo Scientific Alfa Aesar L02731.AC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Widely used hydroxyl-protecting reagent3,4-Dihydro-2H-pyran is used as a hydroxyl-protecting reagent in organic synthesis. It acts as an intermediate in synthetic chemistry. It is used to protect various reactive functional groups. It is involved in the polymerization reaction either alone or with unsaturated compound and finds application in polymer industries. Further, it is employed in the preparation of bicyclic compounds of epoxide-fused, halo compounds and allenic alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Widely used hydroxyl-protecting reagent3,4-Dihydro-2H-pyran is used as a hydroxyl-protecting reagent in organic synthesis. It acts as an intermediate in synthetic chemistry. It is used to protect various reactive functional groups. It is involved in the polymerization reaction either alone or with unsaturated compound and finds application in polymer industries. Further, it is employed in the preparation of bicyclic compounds of epoxide-fused, halo compounds and allenic alcohols.

Solubility
Miscible with water and chloroform.

Notes
Incompatible with oxidizing agents, strong acids and alcohols.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-87-2
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.118
MDL Number MFCD00006558
InChI Key BUDQDWGNQVEFAC-UHFFFAOYSA-N
Synonym dihydropyran, 3,4-dihydropyran, 2,3-dihydropyran, 2h-3,4-dihydropyran, 2,3-dihydro-4h-pyran, dihydro-2h-pyran, 2h-pyran, dihydro, 2h-pyran, 3,4-dihydro, 5,6-dihydro-4h-pyran, dihydropyrane
PubChem CID 8080
IUPAC Name 3,4-dihydro-2H-pyran
SMILES C1CC=COC1

Specifications

Melting Point -70°C
Density 0.926
pH 7.0
Boiling Point 85°C to 86°C
Flash Point −15°C (5°F)
Odor Unpleasant
Refractive Index 1.442
Quantity 25 mL
UN Number UN2376
Beilstein 103493
Solubility Information Miscible with water and chloroform.
Formula Weight 84.12
Percent Purity 99%
Chemical Name or Material 3,4-Dihydro-2H-pyran
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ21Z028 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberL02731.AP,L02731.AC,L02731.AE
Lot NumberA0448722 Certificate TypeCertificate of Analysis Date23/02/2024 Catalog NumberL02731.AP,L02731.AC,L02731.AE
Lot Number10239067 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberL02731.AP,L02731.AC,L02731.AE
Lot Number10190780 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02731.AP,L02731.AC,L02731.AE
Lot Number10218536 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL02731.AP,L02731.AC,L02731.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dihydro-2H-pyran, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.