missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Dihydroxy-3-cyclobutene-1,2-dione, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Description
3,4-Dihydroxy-3-cyclobutene-1,2-dione is used to produce diethoxy-cyclobutenedione by heating, pharmaceutical intermediates, squarylium dyes, and photoconducting squaraines; The dibutylester is used as a contact sensitizer in the treatment of alopecia and warts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-Dihydroxy-3-cyclobutene-1,2-dione is used to produce diethoxy-cyclobutenedione by heating, pharmaceutical intermediates, squarylium dyes, and photoconducting squaraines; The dibutylester is used as a contact sensitizer in the treatment of alopecia and warts.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
3,4-Dihydroxy-3-cyclobutene-1,2-dione is used to produce diethoxy-cyclobutenedione by heating, pharmaceutical intermediates, squarylium dyes, and photoconducting squaraines; The dibutylester is used as a contact sensitizer in the treatment of alopecia and warts.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | >300°C |
Flash Point | 190°C (374°F) |
Quantity | 1 g |
UN Number | UN3261 |
Beilstein | 774275 |
Solubility Information | Soluble in water. |
Formula Weight | 114.06 |
Percent Purity | ≥98% |
Chemical Name or Material | 3,4-Dihydroxy-3-cyclobutene-1,2-dione |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'