missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98% Product Code.: 11364288

Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98%

Product Code. 11364288
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11364288

Brand: Thermo Scientific Chemicals 041535.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,4-Dihydroxy-DL-phenylalanine is used as a dopamine precursor. It is also useful in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63-84-3
Molecular Formula C9H11NO4
Molecular Weight (g/mol) 197.19
MDL Number MFCD00063060
InChI Key WTDRDQBEARUVNC-UHFFFAOYNA-N
Synonym dl-dopa, 3,4-dihydroxy-dl-phenylalanine, 2-amino-3-3,4-dihydroxyphenyl propanoic acid, dl-dioxyphenylalanine, dl-dihydroxyphenylalanine, 3-3,4-dihydroxyphenyl-dl-alanine, 3-hydroxytyrosine, 3-hydroxy-dl-tyrosine, dopa, dl-3,4-dopa
PubChem CID 836
ChEBI CHEBI:49168
IUPAC Name 2-amino-3-(3,4-dihydroxyphenyl)propanoic acid
SMILES NC(CC1=CC=C(O)C(O)=C1)C(O)=O

Specifications

Melting Point 290°C (decomposition)
Merck Index 14,3420
Solubility Information Soluble in water,0.5M dilute hydrochloric acid and formic acid. Insoluble in ethanol,benzene,chloroform and ethyl acetate.
Quantity 5 g
Odor Odorless
Formula Weight 197.19
Sensitivity Air Sensitive
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 3,4-Dihydroxy-DL-phenylalanine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals 3,4-Dihydroxy-DL-phenylalanine, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.