Learn More
Thermo Scientific Chemicals 3,4-Dihydroxy-L-phenylalanine, 98+%
Description
3,4-dihydroxy-L-phenylalanine (DOPA) is used as a cell adhesion molecule in serum-free cultures of anchorage-dependent mammalian cells. It is also used as an anticholinergic drug and used in the treatment of Parkinson's disease. It acts as an intermediate in the biosynthesis of other catecholamines (norepinephrine, epinephrine) and melanin. It acts as a precursor of dopamine product of tyrosine hydroxylase.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- 3-(3,4-Dihydroxyphenyl)-L-alanine, or L-dopa, is converted to dopamine by DOPA decarboxylase in the body
- Upon conversion to dopamine, it acts as an excitatory neurotransmitter
Applications
- L-dopa is useful in the laboratory study of Parkinson's to replenish dopamine in depleted circuits
- L-dopa has also been used for laboratory research on hypertension and melanocyte activity

Specifications
Specifications
Melting Point | ∼295°C (decomposition) |
Beilstein | 2215169 |
Merck Index | 14,5464 |
Solubility Information | Slightly soluble in water,dilute hydrochloric acid and formic acid. Insoluble in ethanol. |
Quantity | 25 g |
Odor | Odorless |
Formula Weight | 197.19 |
Sensitivity | Light sensitive |
Percent Purity | ≥98% |
Chemical Name or Material | 3,4-Dihydroxy-L-phenylalanine |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y15K059 | Certificate of Analysis | 19/11/2024 | A11311.14,A11311.22,A11311.06 |
X21K022 | Certificate of Analysis | 30/10/2024 | A11311.14,A11311.22,A11311.06 |
T29K066 | Certificate of Analysis | 21/08/2024 | A11311.14,A11311.22,A11311.06 |
Q23K033 | Certificate of Analysis | 11/06/2024 | A11311.14,A11311.22,A11311.06 |
S06I024 | Certificate of Analysis | 24/10/2023 | A11311.14,A11311.22,A11311.06 |
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70%
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.