missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 10478060
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-85-5
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.12
MDL Number MFCD00003370
InChI Key IBGBGRVKPALMCQ-UHFFFAOYSA-N
Synonym protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t
PubChem CID 8768
ChEBI CHEBI:50205
IUPAC Name 3,4-dihydroxybenzaldehyde
SMILES C1=CC(=C(C=C1C=O)O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 152.0°C to 157.0°C
Color Beige to Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula (HO)2C6H3CHO
Quantity 25 g
Beilstein 08,246
Merck Index 15,8002
Solubility Information Solubility in water: 50g/L (20°C). Other solubilities: 790g/L ethanol (78°C),freely soluble in ether
Formula Weight 138.12
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 4-Dihydroxybenzaldehyde, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'