Learn More
3,4-Dihydroxybenzaldehyde, 97%, Thermo Scientific Chemicals
£106.00 - £279.00
Chemical Identifiers
CAS | 139-85-5 |
---|---|
Molecular Formula | C7H6O3 |
Molecular Weight (g/mol) | 138.12 |
MDL Number | MFCD00003370 |
InChI Key | IBGBGRVKPALMCQ-UHFFFAOYSA-N |
Synonym | protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t |
PubChem CID | 8768 |
ChEBI | CHEBI:50205 |
IUPAC Name | 3,4-dihydroxybenzaldehyde |
SMILES | C1=CC(=C(C=C1C=O)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10478060
|
Thermo Scientific Acros
114850250 |
25 g | Glass bottle |
£106.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10254570
|
Thermo Scientific Acros
114851000 |
100 g | Glass bottle |
£279.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
139-85-5 | |
138.12 | |
IBGBGRVKPALMCQ-UHFFFAOYSA-N | |
8768 | |
3,4-dihydroxybenzaldehyde |
C7H6O3 | |
MFCD00003370 | |
protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t | |
CHEBI:50205 | |
C1=CC(=C(C=C1C=O)O)O |
Specifications
139-85-5 | |
100.0 | |
Beige to Brown-Gray | |
96% min. (HPLC) | |
(HO)2C6H3CHO | |
08,246 | |
protocatechualdehyde, rancinamycin iv, protocatechuic aldehyde, benzaldehyde, 3,4-dihydroxy, 3,4-dihydroxybenzenecarbonal, 4-formyl-1,2-dihydroxybenzene, 3,4-dihydroxy-benzaldehyde, 4-formyl-1,2-benzenediol, 1,2-dihydroxy-4-formylbenzene, unii-4pvp2hch4t | |
IBGBGRVKPALMCQ-UHFFFAOYSA-N | |
3,4-dihydroxybenzaldehyde | |
8768 | |
138.12 | |
Crystalline Powder |
96.0 | |
152.0°C to 157.0°C | |
Authentic | |
C7H6O3 | |
MFCD00003370 | |
15,8002 | |
Solubility in water: 50g/L (20°C). Other solubilities: 790g/L ethanol (78°C),freely soluble in ether | |
C1=CC(=C(C=C1C=O)O)O | |
138.12 | |
CHEBI:50205 | |
97% | |
3, 4-Dihydroxybenzaldehyde, 97% |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-377-7
missing translation for 'rtecsNumber' : UL0380000
missing translation for 'tsca' : TSCA
RUO – Research Use Only