missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydroxycinnamic acid, predominantly trans, 98+%, Thermo Scientific Chemicals

£58.49 - £190.95

Chemical Identifiers

CAS 331-39-5
Molecular Formula C9H8O4
Molecular Weight (g/mol) 180.159
MDL Number MFCD00004392
InChI Key QAIPRVGONGVQAS-DUXPYHPUSA-N
Synonym caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
PubChem CID 689043
ChEBI CHEBI:16433
IUPAC Name (E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11498496
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A15950.09
10 g
£58.49
10g
Please to purchase this item. Need a web account? Register with us today!
11408506
View Documents
Thermo Scientific Alfa Aesar
A15950.18
50 g
£190.95
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dihydroxycinnamic acid, predominantly trans stimulates prostaglandin synthesis at high doses. It has antioxidant, anti-inflammatory, anti-tumor and immunomodulatory properties.

Solubility
Soluble in DMSO (40 mg/mol), ethanol (25 mg/mL warm), DMF (∼7 mg/mL), PBS, PH 7.2 (∼0.65 mg/mol), hot water, ethyl acetate, Chloroform Hexane, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases and strong oxidizing agents.
Specifications

Chemical Identifiers

331-39-5
180.159
QAIPRVGONGVQAS-DUXPYHPUSA-N
689043
(E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
C9H8O4
MFCD00004392
caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
CHEBI:16433
C1=CC(=C(C=C1C=CC(=O)O)O)O

Specifications

331-39-5
C9H8O4
2210883
caffeic acid, 3,4-dihydroxycinnamic acid, 3-3,4-dihydroxyphenyl acrylic acid, trans-caffeate, 3,4-dihydroxy-trans-cinnamate, 2e-3-3,4-dihydroxyphenyl prop-2-enoic acid, 3,4-dihydroxybenzeneacrylic acid, e-3-3,4-dihydroxyphenyl acrylic acid, trans-caffeic acid, 3-3,4-dihydroxyphenyl propenoic acid
QAIPRVGONGVQAS-DUXPYHPUSA-N
(E)-3-(3,4-dihydroxyphenyl)prop-2-enoic acid
689043
180.16
3,4-Dihydroxycinnamic acid, predominantly trans
∼202°C (decomposition)
MFCD00004392
14,1635
Soluble in DMSO (40mg/mol),ethanol (25mg/mL warm),DMF (∽7mg/mL),PBS,PH 7.2 (∽0.65mg/mol),hot water,ethyl acetate,Chloroform Hexane,and methanol.
C1=CC(=C(C=C1C=CC(=O)O)O)O
180.159
CHEBI:16433
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c

H315-H319-H335-H351-H361

missing translation for 'einecsNumber' : 206-361-2

missing translation for 'rtecsNumber' : GD8950000

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only