missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzhydrazide, 98+%, Thermo Scientific Chemicals

Product Code. 11448593
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41764-74-3
Molecular Formula C9H12N2O3
Molecular Weight (g/mol) 196.206
MDL Number MFCD00017061
InChI Key LJMQIGMMUZLDOC-UHFFFAOYSA-N
PubChem CID 1530859
IUPAC Name 3,4-dimethoxybenzohydrazide
SMILES COC1=C(C=C(C=C1)C(=O)NN)OC

Specifications

Melting Point 143°C to 146°C
Quantity 5 g
Beilstein 2651843
Synonym 3,4-dimethoxybenzhydrazide, veratrohydrazide, 3,4-dimethoxybenzenecarbohydrazide, 3,4-dimethoxybenzoic acid hydrazide, 1,2-dimethoxybenzene-4-carbohydrazide, veratric acid hydrazide, acmc-1aq0x, dimethoxybenzenecarbohydrazide, benzoic acid,3,4-dimethoxy-, hydrazide, benzoic acid, 3,4-dimethoxy-, hydrazide
Formula Weight 196.21
Percent Purity ≥98%
Chemical Name or Material 3,4-Dimethoxybenzhydrazide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'