missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzyl alcohol, 96%, Thermo Scientific Chemicals

Product Code. 10142810
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 93-03-8
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
InChI Key OEGPRYNGFWGMMV-UHFFFAOYSA-N
Synonym 3,4-dimethoxybenzyl alcohol, veratryl alcohol, 3,4-dimethoxyphenyl methanol, benzenemethanol, 3,4-dimethoxy, 3,4-dimethoxyphenylmethyl alcohol, veratrole alcohol, 3,4-dimethoxybenzenemethanol, unii-mb4t4a711h, 3,4-dimethoxyphenyl-methanol
PubChem CID 7118
ChEBI CHEBI:62150
IUPAC Name (3,4-dimethoxyphenyl)methanol
SMILES COC1=C(C=C(C=C1)CO)OC

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 22.0°C
Color Colorless to Yellow
Density 1.1500g/mL
Boiling Point 296.0°C to 297.0°C (732.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH2OH
Refractive Index 1.5510 to 1.5540
Quantity 25 g
Beilstein 06,1113
Specific Gravity 1.15
Solubility Information Solubility in water: miscible
Formula Weight 168.19
Percent Purity 96%
Physical Form Oily Liquid
Chemical Name or Material 3, 4-Dimethoxybenzyl alcohol
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxybenzyl alcohol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dimethoxybenzyl alcohol, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.