missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-1H-pyrazole, 97%, Thermo Scientific Chemicals

Product Code. 15447737
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3,4-Dimethyl-1H-pyrazole is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethyl-1H-pyrazole is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2820-37-3
Molecular Formula C5H8N2
Molecular Weight (g/mol) 96.133
MDL Number MFCD00462154
InChI Key VQTVFIMEENGCJA-UHFFFAOYSA-N
Synonym 3,4-dimethyl-1h-pyrazole, 3 5 ,4-dimethylpyrazole, 3,4-dimethylpyrazole, 3,4-dimethyl pyrazole, 1h-pyrazole, 3,4-dimethyl, unii-7lf1k5quj0, 7lf1k5quj0, 1h-pyrazole,4,5-dimethyl, 1h-pyrazole, 4,5-dimethyl, pyrazole, 3,4-dimethyl
PubChem CID 137735
IUPAC Name 4,5-dimethyl-1H-pyrazole
SMILES CC1=C(NN=C1)C

Specifications

Melting Point 53°C to 55°C
Color Cream
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 96.13
Percent Purity 97%
Physical Form Fused Solid
Chemical Name or Material 3,4-Dimethyl-1H-pyrazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.