missing translation for 'onlineSavingsMsg'
Learn More

3',4'-Dimethylacetanilide, 98+%, Thermo Scientific™

Catalog Number 11331437
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
2198-54-1
C10H13NO
163.22
MFCD00026137
UAOIEEWQVAXCFY-UHFFFAOYSA-N
3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide
75144
N-(3,4-dimethylphenyl)acetamide
CC1=C(C=C(C=C1)NC(=O)C)C
This item is not returnable. View return policy
2198-54-1
C10H13NO
163.22
MFCD00026137
UAOIEEWQVAXCFY-UHFFFAOYSA-N
3',4'-dimethylacetanilide, n-3,4-dimethylphenyl acetamide, 3,4-dimethylacetanilide, n-acetyl-3,4-xylidine, 3',4'-acetoxylidide, acetamide, n-3,4-dimethylphenyl, 3',4'-acetoxylide, acetamide, n-3,4-dimethylphenyl-9ci, 4-acetamino-o-xylol, aceto-3,4-xylidide
75144
N-(3,4-dimethylphenyl)acetamide
CC1=C(C=C(C=C1)NC(=O)C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 96°C to 98°C
Quantity 5 g
Beilstein 387706
Formula Weight 163.22
Percent Purity ≥98%
Chemical Name or Material 3',4'-Dimethylacetanilide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'