missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzenesulfonyl chloride, 98%

Product Code. 11446556
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11446556

Brand: Thermo Scientific Alfa Aesar B24806.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2905-30-8 | C8H9ClO2S | 204.668 g/mol

3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-Dimethylbenzenesulfonyl chloride is used as an intermediate.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agent, bases, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-30-8
Molecular Formula C8H9ClO2S
Molecular Weight (g/mol) 204.668
MDL Number MFCD00625742
InChI Key DNMQPRPJIWTNAX-UHFFFAOYSA-N
Synonym 3,4-dimethylbenzene-1-sulfonyl chloride, 3,4-dimethylbenzenesulfonylchloride, 3,4-dimethyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 3,4-dimethyl, 3,4-dimethylphenyl chlorosulfone, ksc562m4t, 3,4-dimethylphenylsulfonyl chloride, 3,4-dimethylbenzenesulphonyl chloride, 3,4-dimethyl benzenesulfonyl chloride
PubChem CID 3689743
IUPAC Name 3,4-dimethylbenzenesulfonyl chloride
SMILES CC1=C(C=C(C=C1)S(=O)(=O)Cl)C

Specifications

Quantity 5 g
Odor Acrid
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 204.68
Percent Purity 98%
Chemical Name or Material 3,4-Dimethylbenzenesulfonyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.