missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 11944981
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-04-5
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
InChI Key OPVAJFQBSDUNQA-UHFFFAOYSA-N
Synonym benzoic acid, 3,4-dimethyl, 3,4-dimethyl-benzoic acid, 1-carboxy-3,4-dimethylbenzene, o-xylene-4-carboxylic acid, 3,4-dimethylbenzoicacid, pubchem14893, 3,4-dimethyl benzoic acid, asym-o-xylylic acid, acmc-1b5p1, 3, 4 dimethyl benzoic acid
PubChem CID 12073
ChEBI CHEBI:64818
IUPAC Name 3,4-dimethylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)C

Specifications

Melting Point 164.0°C to 168.0°C
CAS Min % 97.5
Color White to Beige
CAS Max % 100.0
Quantity 100 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Powder
Assay Percent Range 97.5% min. (HPLC)
Linear Formula (CH3)2C6H3CO2H
Beilstein 09,II,353
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 3, 4-Dimethylbenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethylbenzoic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'