missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dimethylthieno[2,3-b]thiophene-2-carbaldehyde, 97%, Thermo Scientific™ Product Code.: 10691673

3,4-Dimethylthieno[2,3-b]thiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10691673
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10691673

Brand: Thermo Scientific Maybridge KM03483DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 159709-36-1
Molecular Formula C9H8OS2
Molecular Weight (g/mol) 196.282
MDL Number MFCD00052282
InChI Key WEAXSPMHCXWYPF-UHFFFAOYSA-N
Synonym 3,4-dimethylthieno 2,3-b thiophene-2-carbaldehyde, thieno 2,3-b thiophene-2-carboxaldehyde,3,4-dimethyl, 3,4-dimethyl-2-formylthieno 2,3-b thiophene, 3,4-dimethylthieno 2,3-b thiophene-5-carbaldehyde, 3,4-dimethylthieno 2,3-b thiophene-2-carboxaldehyde, 3,4-dimethylthiopheno 2,3-b thiophene-2-carbaldehydeShow MoreShow Less
PubChem CID 2778771
IUPAC Name 3,4-dimethylthieno[2,3-b]thiophene-5-carbaldehyde
SMILES CC1=CSC2=C1C(=C(S2)C=O)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 196.29
Percent Purity 97%
Chemical Name or Material 3, 4-Dimethylthieno[2, 3-b]thiophene-2-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dimethylthieno[2,3-b]thiophene-2-carbaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.