missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-Epoxytetrahydrofuran, 96%, Thermo Scientific Chemicals
£43.50 - £1001.00
Chemical Identifiers
CAS | 285-69-8 |
---|---|
Molecular Formula | C4H6O2 |
Molecular Weight (g/mol) | 86.09 |
MDL Number | MFCD00800639 |
InChI Key | AIUTZIYTEUMXGG-UHFFFAOYNA-N |
Synonym | 3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran |
PubChem CID | 67511 |
IUPAC Name | 3,6-dioxabicyclo[3.1.0]hexane |
SMILES | C1OCC2OC12 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10429242
|
Thermo Scientific Acros
425530010 |
1 g | Glass bottle |
£43.50
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10041731
|
Thermo Scientific Acros
425530100 |
10 g | Glass bottle |
£213.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11906181
|
Thermo Scientific Acros
425530500 |
50 g | Glass Bottle |
£1001.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
285-69-8 | |
86.09 | |
AIUTZIYTEUMXGG-UHFFFAOYNA-N | |
67511 | |
C1OCC2OC12 |
C4H6O2 | |
MFCD00800639 | |
3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran | |
3,6-dioxabicyclo[3.1.0]hexane |
Specifications
285-69-8 | |
1.2370g/mL | |
Liquid | |
95% min. (GC) | |
C4H6O2 | |
3,4-epoxytetrahydrofuran, 3,6-dioxabicyclo 3.1.0 hexane, 3,6-dioxa-bicyclo 3.1.0 hexane, acmc-1ccip, 3,4-epoxy-tetrahydrofuran, 3,4-diepoxytetrahydrofuran, 3,4-epoxy tetrahydrofuran | |
AIUTZIYTEUMXGG-UHFFFAOYNA-N | |
3,6-dioxabicyclo[3.1.0]hexane | |
67511 | |
96% |
Yellow | |
Authentic | |
1.4450 to 1.4490 | |
1.237 | |
MFCD00800639 | |
Solubility in water: moderately soluble in water. | |
C1OCC2OC12 | |
86.09 | |
86.09 | |
3, 4-Epoxytetrahydrofuran |
Safety and Handling
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 206-006-1
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3,4-Epoxytetrahydrofuran
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only