Learn More
3,4-Ethylenedioxythiophene, 97%, Thermo Scientific Chemicals
£42.21 - £147.74
Chemical Identifiers
CAS | 126213-50-1 |
---|---|
Molecular Formula | C6H6O2S |
Molecular Weight (g/mol) | 142.172 |
MDL Number | MFCD02093622 |
InChI Key | GKWLILHTTGWKLQ-UHFFFAOYSA-N |
Synonym | 3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane |
PubChem CID | 4421864 |
IUPAC Name | 2,3-dihydrothieno[3,4-b][1,4]dioxine |
SMILES | C1COC2=CSC=C2O1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15473018
|
Thermo Scientific Alfa Aesar
H56533.06 |
5 g |
£42.21
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15483018
|
Thermo Scientific Alfa Aesar
H56533.14 |
25 g |
£147.74
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3,4-Ethylenedioxythiophene is used as a monomer to synthesize the conductive polymers and used as a reductant in the one-pot synthesis of gold nanoparticles from chloroauric acid, as starting material used in palladium-catalyzed mono and bis-arylation reactions and in the synthesis of conjugated polymers and copolymers, with potential optical applications. It is also used in redox activity, electroactivity and conductivity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3,4-Ethylenedioxythiophene is used as a monomer to synthesize the conductive polymers and used as a reductant in the one-pot synthesis of gold nanoparticles from chloroauric acid, as starting material used in palladium-catalyzed mono and bis-arylation reactions and in the synthesis of conjugated polymers and copolymers, with potential optical applications. It is also used in redox activity, electroactivity and conductivity.
Solubility
Immsicible with water. Miscible with alcohol and ether.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
126213-50-1 | |
142.172 | |
GKWLILHTTGWKLQ-UHFFFAOYSA-N | |
4421864 | |
C1COC2=CSC=C2O1 |
C6H6O2S | |
MFCD02093622 | |
3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane | |
2,3-dihydrothieno[3,4-b][1,4]dioxine |
Specifications
126213-50-1 | |
193°C | |
C6H6O2S | |
MFCD02093622 | |
3,4-ethylenedioxythiophene, 2,3-dihydrothieno 3,4-b 1,4 dioxine, edot, 3,4-ethylenedioxy thiophene, thieno 3,4-b-1,4-dioxin, 2,3-dihydro, 2,3-dihydrothieno 3,4-b-1,4-dioxin, 2h,3h-thieno 3,4-b 1,4 dioxine, 2h,3h-thiopheno 3,4-e 1,4-dioxane | |
GKWLILHTTGWKLQ-UHFFFAOYSA-N | |
2,3-dihydrothieno[3,4-b][1,4]dioxine | |
4421864 | |
97% |
1.331 | |
104°C (219°F) | |
1.5765 | |
UN2810 | |
Immsicible with water. Miscible with alcohol and ether. | |
C1COC2=CSC=C2O1 | |
142.172 | |
142.17 | |
3,4-Ethylenedioxythiophene |
Sicherheit und Handhabung
GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P361-P363-P501c
H302-H311-H319
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 603-128-0
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only