missing translation for 'onlineSavingsMsg'
Learn More

3-(4-hydroxyphenyl)propanohydrazide, Thermo Scientific™

Product Code. 10506415
Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 65330-63-4
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.21
MDL Number MFCD00025134
InChI Key JMNHUOPRGLTNJI-UHFFFAOYSA-N
Synonym 3-4-hydroxyphenyl propanohydrazide, 3-4-hydroxyphenyl propanehydrazide, 3-4-hydroxyphenyl propionohydrazide, benzenepropanoic acid,4-hydroxy-, hydrazide, benzenepropanoic acid, 4-hydroxy-, hydrazide
PubChem CID 2796587
IUPAC Name 3-(4-hydroxyphenyl)propanehydrazide
SMILES NNC(=O)CCC1=CC=C(O)C=C1

Specifications

Packaging Amber glass bottle
Quantity 5 g
Formula Weight 180.21
Percent Purity 97%
Chemical Name or Material 3-(4-hydroxyphenyl)propanohydrazide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'