missing translation for 'onlineSavingsMsg'
Learn More
3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride, 97%, Thermo Scientific™ Product Code.: 10712873

3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride, 97%, Thermo Scientific™

Product Code. 10712873
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10712873

Brand: Thermo Scientific Maybridge MO00394DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 465514-03-8
Molecular Formula C12H10ClNO3
Molecular Weight (g/mol) 251.67
MDL Number MFCD03407359
InChI Key FABGSPUDPOKWCR-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-5-methyl-4-isoxazolecarbonyl chloride, 3-4-methoxyphenyl-5-methyl-1,2-oxazole-4-carbonyl chloride, 3-4-methoxyphenyl-5-methylisoxazole-4-carbonyl chlorideShow MoreShow Less
PubChem CID 2779766
IUPAC Name 3-(4-methoxyphenyl)-5-methyl-1,2-oxazole-4-carbonyl chloride
SMILES COC1=CC=C(C=C1)C1=NOC(C)=C1C(Cl)=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 251.67
Percent Purity 97%
Chemical Name or Material 3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(4-Methoxyphenyl)-5-methyl-4-isoxazolecarbonyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.