Learn More
3-(4-Methoxyphenyl)propionyl chloride, 99%, Thermo Scientific Chemicals
£63.10 - £249.00
Chemical Identifiers
CAS | 15893-42-2 |
---|---|
Molecular Formula | C10H11ClO2 |
Molecular Weight (g/mol) | 198.65 |
MDL Number | MFCD02258671 |
InChI Key | FQVJPHCAWYRYCK-UHFFFAOYSA-N |
Synonym | 3-4-methoxyphenyl propionyl chloride, 3-4-methoxyphenyl propanoyl chloride, acmc-20anjm, p-methoxyhydrocinnamoyl chloride, 4-methoxyphenylpropionyl chloride, 4-methoxydihydrocinnamoyl chloride, 3-4-methoxyphenyl propanyl chloride, benzenepropanoyl chloride, 4-methoxy, 3-4-methoxyphenyl-propionyl chloride, 3-4-methoxy-phenyl-propionyl chloride |
PubChem CID | 10932441 |
IUPAC Name | 3-(4-methoxyphenyl)propanoyl chloride |
SMILES | COC1=CC=C(CCC(Cl)=O)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355429
|
Thermo Scientific Alfa Aesar
H50416.03 |
1 g |
£63.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365429
|
Thermo Scientific Alfa Aesar
H50416.06 |
5 g |
£249.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(4-Methoxyphenyl)propionyl chloride is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture. Incompatible with bases.
Chemical Identifiers
15893-42-2 | |
198.65 | |
FQVJPHCAWYRYCK-UHFFFAOYSA-N | |
10932441 | |
COC1=CC=C(CCC(Cl)=O)C=C1 |
C10H11ClO2 | |
MFCD02258671 | |
3-4-methoxyphenyl propionyl chloride, 3-4-methoxyphenyl propanoyl chloride, acmc-20anjm, p-methoxyhydrocinnamoyl chloride, 4-methoxyphenylpropionyl chloride, 4-methoxydihydrocinnamoyl chloride, 3-4-methoxyphenyl propanyl chloride, benzenepropanoyl chloride, 4-methoxy, 3-4-methoxyphenyl-propionyl chloride, 3-4-methoxy-phenyl-propionyl chloride | |
3-(4-methoxyphenyl)propanoyl chloride |
Specifications
15893-42-2 | |
MFCD02258671 | |
Air and moisture sensitive | |
Reacts with water. | |
COC1=CC=C(CCC(Cl)=O)C=C1 | |
198.65 | |
198.65 | |
3-(4-Methoxyphenyl)propionyl chloride |
C10H11ClO2 | |
UN3265 | |
3-4-methoxyphenyl propionyl chloride, 3-4-methoxyphenyl propanoyl chloride, acmc-20anjm, p-methoxyhydrocinnamoyl chloride, 4-methoxyphenylpropionyl chloride, 4-methoxydihydrocinnamoyl chloride, 3-4-methoxyphenyl propanyl chloride, benzenepropanoyl chloride, 4-methoxy, 3-4-methoxyphenyl-propionyl chloride, 3-4-methoxy-phenyl-propionyl chloride | |
FQVJPHCAWYRYCK-UHFFFAOYSA-N | |
3-(4-methoxyphenyl)propanoyl chloride | |
10932441 | |
99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only