Learn More
3-(4-Methylbenzoyl)propionic acid, 98%, Thermo Scientific Chemicals
£32.80 - £466.32
Chemical Identifiers
CAS | 4619-20-9 |
---|---|
Molecular Formula | C11H12O3 |
Molecular Weight (g/mol) | 192.214 |
MDL Number | MFCD00020541 |
InChI Key | OEEUWZITKKSXAZ-UHFFFAOYSA-N |
Synonym | 3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo |
PubChem CID | 244162 |
IUPAC Name | 4-(4-methylphenyl)-4-oxobutanoic acid |
SMILES | CC1=CC=C(C=C1)C(=O)CCC(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494023
|
Thermo Scientific Alfa Aesar
A10290.06 |
5 g |
£32.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404033
|
Thermo Scientific Alfa Aesar
A10290.14 |
25 g |
£123.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414033
|
Thermo Scientific Alfa Aesar
A10290.22 |
100 g |
£466.32
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-(4-Methylbenzoyl)propionic acid can be used as an intermediate in agrochemical, pharmaceutical and dyestuff field
Solubility
Insoluble in water.
Notes
Incompatible with bases and oxidizing agents. Store in a cool, dry condition in well sealed containers.
Chemical Identifiers
4619-20-9 | |
192.214 | |
OEEUWZITKKSXAZ-UHFFFAOYSA-N | |
244162 | |
CC1=CC=C(C=C1)C(=O)CCC(=O)O |
C11H12O3 | |
MFCD00020541 | |
3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo | |
4-(4-methylphenyl)-4-oxobutanoic acid |
Specifications
4619-20-9 | |
C11H12O3 | |
2099653 | |
Insoluble in water. | |
CC1=CC=C(C=C1)C(=O)CCC(=O)O | |
192.214 | |
192.22 | |
3- (4-Methylbenzoyl)propionic acid |
124°C to 128°C | |
MFCD00020541 | |
3-4-methylbenzoyl propionic acid, 4-oxo-4-p-tolyl butanoic acid, 4-4-methylphenyl-4-oxobutanoic acid, 4-4-methylphenyl-4-oxobutyric acid, 4-oxo-4-4-tolyl butyric acid, 4-oxo-4-p-tolylbutyric acid, 4-oxo-4-p-tolylbutanoic acid, 3-4-methylbenzoyl-propionic acid, acmc-1aooh, benzenebutanoic acid, 4-methyl-gamma-oxo | |
OEEUWZITKKSXAZ-UHFFFAOYSA-N | |
4-(4-methylphenyl)-4-oxobutanoic acid | |
244162 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.