missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole, Alfa Aesar™
£60.58 - £217.00
Chemical Identifiers
CAS | 338393-13-8 |
---|---|
Molecular Formula | C10H11N3O2S |
Molecular Weight (g/mol) | 237.277 |
MDL Number | MFCD01568307 |
InChI Key | YYQTWXHTIYSIRV-UHFFFAOYSA-N |
Synonym | 3-4-methylbenzyl sulfonyl-1h-1,2,4-triazole, 4-methylbenzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methanesulfonyl-1h-1,2,4-triazole, acmc-20anb6, methylbenzyltriazolylsulfone, 3-p-tolylmethylsulfonyl-1h-1,2,4-triazole, 3-p-tolylmethanesulfonyl-1h-1,2,4 triazole, 4-methyl-benzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methyl sulfonyl-1,2,4-triazole |
PubChem CID | 2063879 |
IUPAC Name | 5-[(4-methylphenyl)methylsulfonyl]-1H-1,2,4-triazole |
SMILES | CC1=CC=C(C=C1)CS(=O)(=O)C2=NC=NN2 |
Description
Chemical Identifiers
338393-13-8 | |
237.277 | |
YYQTWXHTIYSIRV-UHFFFAOYSA-N | |
2063879 | |
CC1=CC=C(C=C1)CS(=O)(=O)C2=NC=NN2 |
C10H11N3O2S | |
MFCD01568307 | |
3-4-methylbenzyl sulfonyl-1h-1,2,4-triazole, 4-methylbenzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methanesulfonyl-1h-1,2,4-triazole, acmc-20anb6, methylbenzyltriazolylsulfone, 3-p-tolylmethylsulfonyl-1h-1,2,4-triazole, 3-p-tolylmethanesulfonyl-1h-1,2,4 triazole, 4-methyl-benzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methyl sulfonyl-1,2,4-triazole | |
5-[(4-methylphenyl)methylsulfonyl]-1H-1,2,4-triazole |
Specifications
338393-13-8 | |
MFCD01568307 | |
YYQTWXHTIYSIRV-UHFFFAOYSA-N | |
5-[(4-methylphenyl)methylsulfonyl]-1H-1,2,4-triazole | |
2063879 | |
3-[(4-Methylbenzyl)sulfonyl]-1H-1,2,4-triazole |
C10H11N3O2S | |
3-4-methylbenzyl sulfonyl-1h-1,2,4-triazole, 4-methylbenzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methanesulfonyl-1h-1,2,4-triazole, acmc-20anb6, methylbenzyltriazolylsulfone, 3-p-tolylmethylsulfonyl-1h-1,2,4-triazole, 3-p-tolylmethanesulfonyl-1h-1,2,4 triazole, 4-methyl-benzyl 1h-1,2,4-triazol-3-yl sulfone, 3-4-methylphenyl methyl sulfonyl-1,2,4-triazole | |
CC1=CC=C(C=C1)CS(=O)(=O)C2=NC=NN2 | |
237.277 | |
237.28 |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title