missing translation for 'onlineSavingsMsg'
Learn More
3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals Product Code.: 11440144

3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11440144
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11440144

Brand: Thermo Scientific Alfa Aesar A13597.30

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3?,4?-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,4′-(Methylenedioxy)acetophenone is used in the synthesis of N-ethyl-1-(3,4-methylenedioxyphenyl)ethylamine.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3162-29-6
Molecular Formula C9H8O3
Molecular Weight (g/mol) 164.16
MDL Number MFCD00005831
InChI Key BMHMKWXYXFBWMI-UHFFFAOYSA-N
Synonym 3',4'-methylenedioxy acetophenone, 1-benzo d 1,3 dioxol-5-yl ethanone, 5-acetyl-1,3-benzodioxole, 3,4-methylenedioxyacetophenone, 3,4-methylenedioxy acetophenone, 3',4'-methylenedioxyacetophenone, 1-1,3-benzodioxol-5-yl ethanone, acetopiperone, 1-1,3-benzodioxol-5-yl ethan-1-one, ethanone, 1-1,3-benzodioxol-5-yl
PubChem CID 76622
IUPAC Name 1-(1,3-benzodioxol-5-yl)ethanone
SMILES CC(=O)C1=CC2=C(C=C1)OCO2

Specifications

Melting Point 85°C to 89°C
Boiling Point 152°C to 155°C (15 mmHg)
Quantity 250 g
Beilstein 139669
Solubility Information Slightly soluble in water.
Formula Weight 164.16
Percent Purity 98%
Chemical Name or Material 3',4'-(Methylenedioxy)acetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3',4'-(Methylenedioxy)acetophenone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.