missing translation for 'onlineSavingsMsg'
Learn More
3,4-(Methylenedioxy)aniline, 97%, Thermo Scientific Chemicals Product Code.: 10581811

3,4-(Methylenedioxy)aniline, 97%, Thermo Scientific Chemicals

Product Code. 10581811
50 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10581811

Brand: Thermo Scientific Acros 171470500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14268-66-7
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.14
MDL Number MFCD00005832
InChI Key XGNXYCFREOZBOL-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy aniline, benzo d 1,3 dioxol-5-amine, 5-amino-1,3-benzodioxole, 3,4-methylenedioxyaniline, 4-amino-1,2-methylenedioxybenzene, 5-aminobenzodioxole, methylenedioxyaniline, 2h-1,3-benzodioxol-5-amine, 1-amino-3,4-methylenedioxybenzene, 1-amino-3,4-dimethylenedioxybenzeneShow MoreShow Less
PubChem CID 84310
IUPAC Name 1,3-benzodioxol-5-amine
SMILES C1OC2=C(O1)C=C(C=C2)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 38.0°C to 43.0°C
Color Black to Brown
Boiling Point 144.0°C (16.0 mmHg)
Flash Point 142°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 50 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 137.14
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 3, 4-(Methylenedioxy)aniline, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443469 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number171470500,171470100
Lot NumberA0431501 Certificate TypeCertificate of Analysis Date22/12/2023 Catalog Number171470500,171470100
Lot NumberA0395305 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number171470500,171470100
Lot NumberA0418392 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number171470500,171470100
Lot NumberA0443469 Certificate TypeCertificate of Analysis Date15/07/2022 Catalog Number171470500,171470100
compliance-icons
Product Identifier
  • 3,4-(Methylenedioxy)aniline
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-(Methylenedioxy)aniline, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.