missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-(Methylenedioxy)benzylideneacetone, 98%, Thermo Scientific Chemicals
£47.60 - £180.00
Chemical Identifiers
CAS | 3160-37-0 |
---|---|
Molecular Formula | C11H10O3 |
Molecular Weight (g/mol) | 190.198 |
MDL Number | MFCD00016907 |
InChI Key | XIYPXOFSURQTTJ-NSCUHMNNSA-N |
Synonym | piperonalacetone, heliotropyl acetone, acetone, piperonylidene, piperonylideneacetone, piperonylidene acetone, unii-t43byj64eg, 3-buten-2-one, 4-1,3-benzodioxol-5-yl, ccris 6270, 3,4-methylenedioxy benzalacetone, 3,4-methylenedioxy benzylideneacetone |
PubChem CID | 6040503 |
IUPAC Name | (E)-4-(1,3-benzodioxol-5-yl)but-3-en-2-one |
SMILES | CC(=O)C=CC1=CC2=C(C=C1)OCO2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382646
|
Thermo Scientific Alfa Aesar
L08675.09 |
10 g |
£47.60
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392646
|
Thermo Scientific Alfa Aesar
L08675.18 |
50 g |
£180.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3160-37-0 | |
190.198 | |
XIYPXOFSURQTTJ-NSCUHMNNSA-N | |
6040503 | |
CC(=O)C=CC1=CC2=C(C=C1)OCO2 |
C11H10O3 | |
MFCD00016907 | |
piperonalacetone, heliotropyl acetone, acetone, piperonylidene, piperonylideneacetone, piperonylidene acetone, unii-t43byj64eg, 3-buten-2-one, 4-1,3-benzodioxol-5-yl, ccris 6270, 3,4-methylenedioxy benzalacetone, 3,4-methylenedioxy benzylideneacetone | |
(E)-4-(1,3-benzodioxol-5-yl)but-3-en-2-one |
Specifications
3160-37-0 | |
C11H10O3 | |
83983 | |
XIYPXOFSURQTTJ-NSCUHMNNSA-N | |
(E)-4-(1,3-benzodioxol-5-yl)but-3-en-2-one | |
6040503 | |
98% |
108°C to 110°C | |
MFCD00016907 | |
piperonalacetone, heliotropyl acetone, acetone, piperonylidene, piperonylideneacetone, piperonylidene acetone, unii-t43byj64eg, 3-buten-2-one, 4-1,3-benzodioxol-5-yl, ccris 6270, 3,4-methylenedioxy benzalacetone, 3,4-methylenedioxy benzylideneacetone | |
CC(=O)C=CC1=CC2=C(C=C1)OCO2 | |
190.198 | |
190.2 | |
3,4-(Methylenedioxy)benzylideneacetone |
Safety and Handling
missing translation for 'einecsNumber' : 221-608-4
missing translation for 'rtecsNumber' : EM9822500
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only