missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,4-(Methylenedioxy)phenylacetic acid, 98%
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Description
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.
Solubility
Soluble in water (partly), ethanol, and methanol.
Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.
3,4-(Methylenedioxy)phenylacetic acid is used as organic intermediates.
Solubility
Soluble in water (partly), ethanol, and methanol.
Notes
Store at room temperature. Store away from oxidizing agents. Store in a cool, dry place. Store in a tightly closed container.
Specifications
Specifica
| Melting Point | 127°C to 130°C |
| Quantity | 100 g |
| Beilstein | 177739 |
| Solubility Information | Soluble in water (partly),ethanol,and methanol. |
| Formula Weight | 180.16 |
| Percent Purity | 98% |
| Chemical Name or Material | 3,4- (Methylenedioxy)phenylacetic acid |
RUO – Research Use Only
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto