missing translation for 'onlineSavingsMsg'
Learn More
3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals Product Code.: 11397466

3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals

Product Code. 11397466
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11397466

Brand: Thermo Scientific Alfa Aesar L09518.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4-(Methylenedioxy)phenylacetonitrile was used in synthesis of derrubone. It was used as standard to analyze the seized methamphetamine samples showing unique profiles of stable isotopic compositions by isotope ratio mass spectrometry.

Solubility
Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4439-02-5
Molecular Formula C9H7NO2
Molecular Weight (g/mol) 161.16
MDL Number MFCD00005835
InChI Key ZQPBOYASBNAXOZ-UHFFFAOYSA-N
Synonym 3,4-methylenedioxy phenylacetonitrile, 1,3-benzodioxole-5-acetonitrile, 3,4-methylenedioxyphenylacetonitrile, 2-benzo d 1,3 dioxol-5-yl acetonitrile, 3,4-methylenedioxybenzyl cyanide, 2-2h-1,3-benzodioxol-5-yl acetonitrile, 1,3-benzodioxol-5-ylacetonitrile, 2-1,3-benzodioxol-5-yl acetonitrile, 3,4-methylenedioxybenzylcyanide, homopiperonylonitrileShow MoreShow Less
PubChem CID 78178
IUPAC Name 2-(1,3-benzodioxol-5-yl)acetonitrile
SMILES C1OC2=C(O1)C=C(C=C2)CC#N

Specifications

Melting Point 41°C to 45°C
Boiling Point 135°C to 140°C (5 mmHg)
Flash Point >110°C (230°F)
Quantity 10 g
UN Number UN3439
Beilstein 7739
Solubility Information Insoluble in water.
Formula Weight 161.16
Percent Purity ≥98%
Chemical Name or Material 3,4-(Methylenedioxy)phenylacetonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10138939 Certificate TypeCertificate of Analysis Date08/11/2023 Catalog NumberL09518.18,L09518.09
Lot Number10138939 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL09518.18,L09518.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-(Methylenedioxy)phenylacetonitrile, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.