missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(4-Morpholinyl)propylamine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
3-(4-Morpholinyl)propylamine is catalytic agent, petrochemical additive. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Fully miscible in water.
Notes
Air Sensitive. Store under dry inert gas. Store away from air. Incompatible with acids, acid chlorides, acid anhydrides.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -15°C |
Density | 0.987 |
Boiling Point | 223°C to 225°C |
Flash Point | 98°C (210°F) |
Refractive Index | 1.4761 |
Quantity | 50 g |
UN Number | UN2735 |
Sensitivity | Air Sensitive |
Solubility Information | Fully miscible in water. |
Formula Weight | 144.22 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'