missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Pyridyl)propionic acid, 97%

Product Code. 15454888
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15454888

Brand: Thermo Scientific Alfa Aesar H58655.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6318-43-0 | C8H9NO2 | 151.165 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6318-43-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00084846
InChI Key WSXGQYDHJZKQQB-UHFFFAOYSA-N
Synonym 3-pyridin-4-yl propanoic acid, 3-pyridin-4-yl-propionic acid, 4-pyridinepropanoic acid, 4-pyridinepropionic acid, 3-4-pyridinyl propanoic acid, 3-4-pyridyl propanoic acid, 3-4-pyridyl propionic acid, 3-pyridin-4-yl propionic acid, 4-carboxyethylpyridine, acmc-1b6yz
PubChem CID 233243
IUPAC Name 3-pyridin-4-ylpropanoic acid
SMILES C1=CN=CC=C1CCC(=O)O

Specifications

Melting Point 231°C
Quantity 1 g
Beilstein 122234
Formula Weight 151.16
Percent Purity 97%
Chemical Name or Material 3-(4-Pyridyl)propionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.