Learn More
3-[4-(Trifluoromethoxy)phenyl]acrylic acid, 97%, Thermo Scientific™
£19.60 - £332.00
Chemical Identifiers
CAS | 783-13-1 |
---|---|
Molecular Formula | C10H7F3O3 |
Molecular Weight (g/mol) | 232.158 |
MDL Number | MFCD00066338 |
InChI Key | RNYVTJANWYBGPW-ZZXKWVIFSA-N |
Synonym | 3-4-trifluoromethoxy phenyl acrylic acid, 4-trifluoromethoxy cinnamic acid, e-3-4-trifluoromethoxy phenyl acrylic acid, e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 2e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-4-trifluoromethoxyphenyl acrylic acid, trifluoromethoxyphenylacrylicacid, trans-4-trifluoromethoxy cinnamic acid |
PubChem CID | 735857 |
IUPAC Name | (E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoic acid |
SMILES | C1=CC(=CC=C1C=CC(=O)O)OC(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10087402
|
Thermo Scientific Maybridge
SEW04310DA |
1 g |
£19.60
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10757183
|
Thermo Scientific Maybridge
SEW04310EA |
10 g |
£112.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10371843
|
Thermo Scientific Maybridge
SEW04310EE |
50 g |
£332.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
783-13-1 | |
232.158 | |
RNYVTJANWYBGPW-ZZXKWVIFSA-N | |
735857 | |
C1=CC(=CC=C1C=CC(=O)O)OC(F)(F)F |
C10H7F3O3 | |
MFCD00066338 | |
3-4-trifluoromethoxy phenyl acrylic acid, 4-trifluoromethoxy cinnamic acid, e-3-4-trifluoromethoxy phenyl acrylic acid, e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 2e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-4-trifluoromethoxyphenyl acrylic acid, trifluoromethoxyphenylacrylicacid, trans-4-trifluoromethoxy cinnamic acid | |
(E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoic acid |
Specifications
783-13-1 | |
97% | |
C10H7F3O3 | |
3-4-trifluoromethoxy phenyl acrylic acid, 4-trifluoromethoxy cinnamic acid, e-3-4-trifluoromethoxy phenyl acrylic acid, e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 2e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-4-trifluoromethoxyphenyl acrylic acid, trifluoromethoxyphenylacrylicacid, trans-4-trifluoromethoxy cinnamic acid | |
C1=CC(=CC=C1C=CC(=O)O)OC(F)(F)F | |
232.158 | |
232.16 | |
3-[4-(Trifluoromethoxy)phenyl]acrylic acid |
97.0 | |
Amber glass bottle | |
MFCD00066338 | |
RNYVTJANWYBGPW-ZZXKWVIFSA-N | |
(E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoic acid | |
735857 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.