missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',5,7-Trihydroxy-4'-methoxyflavanone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.
Solubility
Soluble in water (partly), dilute alkalis, and ethanol (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.
Solubility
Soluble in water (partly), dilute alkalis, and ethanol (50 mg/ml).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 227°C to 232°C |
Quantity | 5 g |
Beilstein | 309850 |
Merck Index | 14,4670 |
Solubility Information | Soluble in water (partly),dilute alkalis,and ethanol (50mg/ml). |
Formula Weight | 302.3 |
Percent Purity | 97% |
Chemical Name or Material | 3',5,7-Trihydroxy-4'-methoxyflavanone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'