missing translation for 'onlineSavingsMsg'
Learn More

3',5'-Bis(trifluoromethyl)acetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11404133
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30071-93-3
Molecular Formula C10H6F6O
Molecular Weight (g/mol) 256.15
MDL Number MFCD00009910
InChI Key MCYCSIKSZLARBD-UHFFFAOYSA-N
Synonym 3',5'-bis trifluoromethyl acetophenone, 3,5-bis trifluoromethyl acetophenone, 1-3,5-bis trifluoromethyl phenyl ethanone, 1-3,5-bis trifluoromethyl phenyl ethan-1-one, mbt-ac, 1-3,5-di trifluoromethyl phenyl ethan-1-one, 3,5-ditrifluoromethylacetophenone, 3',5'-bis-trifluoromethylacetophenone
PubChem CID 121616
SMILES CC(=O)C1=CC(=CC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Density 1.422
Boiling Point 85°C (8 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.423
Quantity 25 g
Beilstein 2384789
IUPAC Name 1-[3,5-bis(trifluoromethyl)phenyl]ethanone
Formula Weight 256.15
Percent Purity 98%
Chemical Name or Material 3',5'-Bis (trifluoromethyl)acetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'